Assay Details
| Gene Symbol: LOC4335606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778870.2 | XP_015634356.1 | 6 - 7 | 1366 | 70 | |
| GenBank mRNA | AK101769.1 | - | 6 - 7 | 1373 | 70 | |
More Information
Set Membership: |
|

LOC4335606
Genomic Map