Assay Details
| Gene Symbol: LOC4335937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781506.2 | XP_015636992.1 | 3 - 4 | 619 | 69 | |
| GenBank mRNA | AB249661.1 | - | 3 - 4 | 255 | 69 | |
| AK072736.1 | - | 3 - 4 | 619 | 69 | ||
| AK105815.1 | - | 3 - 4 | 421 | 69 | ||
More Information
Set Membership: |
|

LOC4335937
Genomic Map