Assay Details
| Gene Symbol: LOC4335937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781506.2 | XP_015636992.1 | 4 - 5 | 689 | 75 | |
| GenBank mRNA | AB249661.1 | - | 4 - 5 | 325 | 75 | |
| AK072736.1 | - | 4 - 5 | 689 | 75 | ||
| AK105815.1 | - | 4 - 5 | 491 | 75 | ||
More Information
Set Membership: |
|

LOC4335937
Genomic Map