Assay Details
| Gene Symbol: LOC4336259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780956.2 | XP_015636442.1 | 3 - 4 | 1087 | 85 | |
| XM_015780957.1 | XP_015636443.1 | 3 - 4 | 851 | 85 | ||
| GenBank mRNA | AK102780.1 | - | 3 - 4 | 1049 | 85 | |
| AK120369.1 | - | 2 - 3 | 786 | 85 | ||
More Information
Set Membership: |
|

LOC4336259
Genomic Map