Assay Details
| Gene Symbol: LOC4336287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781765.2 | XP_015637251.1 | 2 - 3 | 236 | 82 | |
| GenBank mRNA | AK059415.1 | - | 2 - 3 | 172 | 82 | |
| AK099054.1 | - | 2 - 3 | 173 | 82 | ||
More Information
Set Membership: |
|

LOC4336287
Genomic Map