Assay Details
| Gene Symbol: LOC4336287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781765.2 | XP_015637251.1 | 4 - 5 | 539 | 114 | |
| GenBank mRNA | AK059415.1 | - | 4 - 5 | 475 | 114 | |
| AK099054.1 | - | 4 - 5 | 476 | 114 | ||
More Information
Set Membership: |
|

LOC4336287
Genomic Map