Assay Details
| Gene Symbol: LOC4336287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781765.2 | XP_015637251.1 | 5 - 6 | 682 | 149 | |
| GenBank mRNA | AK059415.1 | - | 5 - 6 | 618 | 149 | |
| AK099054.1 | - | 5 - 6 | 619 | 149 | ||
More Information
Set Membership: |
|

LOC4336287
Genomic Map