Assay Details
| Gene Symbol: LOC4336287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781765.2 | XP_015637251.1 | 8 - 9 | 1212 | 73 | |
| GenBank mRNA | AK058436.1 | - | 406 | 73 | ||
| AK059415.1 | - | 8 - 9 | 1148 | 73 | ||
| AK099054.1 | - | 8 - 9 | 1149 | 73 | ||
More Information
Set Membership: |
|

LOC4336287
Genomic Map