Assay Details
| Gene Symbol: LOC4336287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781765.2 | XP_015637251.1 | 9 - 10 | 1293 | 72 | |
| GenBank mRNA | AK058436.1 | - | 487 | 72 | ||
| AK059415.1 | - | 9 - 10 | 1229 | 72 | ||
| AK099054.1 | - | 9 - 10 | 1230 | 72 | ||
More Information
Set Membership: |
|

LOC4336287
Genomic Map