Assay Details
| Gene Symbol: LOC4336308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778481.2 | XP_015633967.1 | 2 - 3 | 748 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4336308| Gene Symbol: LOC4336308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778481.2 | XP_015633967.1 | 2 - 3 | 748 | 68 | |
Set Membership: |
|