Assay Details
| Gene Symbol: LOC4336345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781188.2 | XP_015636674.1 | 14 - 15 | 1576 | 134 | |
| XM_015781189.2 | XP_015636675.1 | 14 - 15 | 1510 | 134 | ||
| GenBank mRNA | AK067042.1 | - | 14 - 15 | 1563 | 134 | |
More Information
Set Membership: |
|

LOC4336345
Genomic Map