Assay Details
| Gene Symbol: LOC4336345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781188.2 | XP_015636674.1 | 21 - 22 | 3410 | 86 | |
| XM_015781189.2 | XP_015636675.1 | 21 - 22 | 3344 | 86 | ||
| GenBank mRNA | AK067042.1 | - | 21 - 22 | 3397 | 86 | |
| BT129098.1 | - | 1 - 2 | 425 | 86 | ||
More Information
Set Membership: |
|

LOC4336345
Genomic Map