Assay Details
| Gene Symbol: LOC4336487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781769.1 | XP_015637255.1 | 4 - 5 | 609 | 74 | |
| GenBank mRNA | AK063584.1 | - | 4 - 5 | 468 | 74 | |
More Information
Set Membership: |
|

LOC4336487
Genomic Map