Assay Details
| Gene Symbol: LOC4336555 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777927.2 | XP_015633413.1 | 1 | 2582 | 88 | |
| GenBank mRNA | AK066297.1 | - | 1 | 2543 | 88 | |
More Information
Set Membership: |
|

LOC4336555
Genomic Map