Assay Details
| Gene Symbol: LOC4336702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780528.2 | XP_015636014.1 | 12 - 13 | 1917 | 115 | |
| XM_015780529.2 | XP_015636015.1 | 12 - 13 | 1917 | 115 | ||
| GenBank mRNA | AK106571.1 | - | 4 - 5 | 1555 | 115 | |
More Information
Set Membership: |
|

LOC4336702
Genomic Map