Assay Details
| Gene Symbol: LOC4336917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781094.2 | XP_015636580.1 | 10 - 11 | 1308 | 108 | |
| GenBank mRNA | AK060852.1 | - | 8 - 9 | 810 | 108 | |
| AK102588.1 | - | 10 - 11 | 1286 | 108 | ||
More Information
Set Membership: |
|

LOC4336917
Genomic Map