Assay Details
| Gene Symbol: LOC4336917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781094.2 | XP_015636580.1 | 13 - 14 | 1660 | 82 | |
| GenBank mRNA | AK060852.1 | - | 11 - 12 | 1162 | 82 | |
More Information
Set Membership: |
|

LOC4336917
Genomic Map