Assay Details
Gene Symbol: LOC4336981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780217.2 | XP_015635703.1 | 2 - 3 | 286 | 107 | |
XM_015780218.2 | XP_015635704.1 | 2 - 3 | 1370 | 107 | ||
XM_015780219.2 | XP_015635705.1 | 1 - 2 | 235 | 107 | ||
GenBank mRNA | AY177698.1 | - | 1 - 2 | 185 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
