Assay Details
| Gene Symbol: LOC4336981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780217.2 | XP_015635703.1 | 3 - 4 | 361 | 91 | |
| XM_015780218.2 | XP_015635704.1 | 3 - 4 | 1445 | 91 | ||
| XM_015780219.2 | XP_015635705.1 | 2 - 3 | 310 | 91 | ||
| GenBank mRNA | AY177698.1 | - | 2 - 3 | 260 | 91 | |
More Information
Set Membership: |
|

LOC4336981
Genomic Map