Assay Details
Gene Symbol: LOC4337137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780183.2 | XP_015635669.1 | 9 - 10 | 1081 | 72 | |
GenBank mRNA | AK070546.1 | - | 9 - 10 | 1071 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
