Assay Details
| Gene Symbol: LOC4337165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780882.1 | XP_015636368.1 | 1 - 2 | 394 | 72 | |
| GenBank mRNA | AB188836.1 | - | 1 - 2 | 188 | 72 | |
| AK070318.1 | - | 1 - 2 | 366 | 72 | ||
More Information
Set Membership: |
|

LOC4337165
Genomic Map