Assay Details
| Gene Symbol: LOC4337165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780882.1 | XP_015636368.1 | 8 - 9 | 945 | 148 | |
| GenBank mRNA | AB188836.1 | - | 8 - 9 | 739 | 148 | |
| AK067948.1 | - | 4 - 5 | 1795 | 148 | ||
| AK070318.1 | - | 7 - 8 | 834 | 148 | ||
More Information
Set Membership: |
|

LOC4337165
Genomic Map