Assay Details
| Gene Symbol: LOC4337165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780882.1 | XP_015636368.1 | 9 - 10 | 1018 | 138 | |
| GenBank mRNA | AK067948.1 | - | 5 - 6 | 1868 | 138 | |
| AK070318.1 | - | 8 - 9 | 907 | 138 | ||
More Information
Set Membership: |
|

LOC4337165
Genomic Map