Assay Details
| Gene Symbol: LOC9269072 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781362.2 | XP_015636848.1 | 8 - 9 | 1587 | 88 | |
| XM_015781363.2 | XP_015636849.1 | 8 - 9 | 1589 | 88 | ||
| GenBank mRNA | AK121346.1 | - | 2 - 3 | 528 | 88 | |
More Information
Set Membership: |
|

LOC9269072
Genomic Map