Assay Details
| Gene Symbol: LOC9269072 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781362.2 | XP_015636848.1 | 9 - 10 | 1673 | 61 | |
| XM_015781363.2 | XP_015636849.1 | 9 - 10 | 1675 | 61 | ||
| GenBank mRNA | AK121346.1 | - | 3 - 4 | 614 | 61 | |
More Information
Set Membership: |
|

LOC9269072
Genomic Map