Assay Details
| Gene Symbol: LOC4337313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781342.2 | XP_015636828.1 | 5 - 6 | 1706 | 66 | |
| GenBank mRNA | AK243269.1 | - | 5 - 6 | 1497 | 66 | |
| AY578162.1 | - | 6 - 7 | 1373 | 66 | ||
More Information
Set Membership: |
|

LOC4337313
Genomic Map