Assay Details
| Gene Symbol: LOC4337737 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK101387.1 | - | 5 - 6 | 1072 | 139 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4337737| Gene Symbol: LOC4337737 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK101387.1 | - | 5 - 6 | 1072 | 139 | |
Set Membership: |
|