Assay Details
| Gene Symbol: LOC4337785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784663.2 | XP_015640149.1 | 10 - 11 | 1550 | 137 | |
| XM_015784664.1 | XP_015640150.1 | 10 - 11 | 1179 | 137 | ||
| GenBank mRNA | AK120211.1 | - | 10 - 11 | 1186 | 137 | |
More Information
Set Membership: |
|

LOC4337785
Genomic Map