Assay Details
| Gene Symbol: LOC4337785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784663.2 | XP_015640149.1 | 15 - 16 | 2437 | 76 | |
| XM_015784664.1 | XP_015640150.1 | 15 - 16 | 2042 | 76 | ||
| GenBank mRNA | AK120211.1 | - | 15 - 16 | 2049 | 76 | |
More Information
Set Membership: |
|

LOC4337785
Genomic Map