Assay Details
| Gene Symbol: LOC4337785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784663.2 | XP_015640149.1 | 9 - 10 | 1385 | 66 | |
| XM_015784664.1 | XP_015640150.1 | 9 - 10 | 1014 | 66 | ||
| GenBank mRNA | AK120211.1 | - | 9 - 10 | 1021 | 66 | |
More Information
Set Membership: |
|

LOC4337785
Genomic Map