Assay Details
| Gene Symbol: LOC4337792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784378.2 | XP_015639864.1 | 1 - 2 | 469 | 112 | |
| GenBank mRNA | AK243398.1 | - | 1 - 2 | 427 | 112 | |
| AK288941.1 | - | 1 - 2 | 476 | 112 | ||
More Information
Set Membership: |
|

LOC4337792
Genomic Map