Assay Details
Gene Symbol: LOC4337931 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015783867.2 | XP_015639353.1 | 2 - 3 | 707 | 100 | |
GenBank mRNA | AK073439.1 | - | 3 - 4 | 498 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
