Assay Details
| Gene Symbol: LOC4338177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782132.2 | XP_015637618.1 | 7 - 8 | 1065 | 66 | |
| XM_015782133.2 | XP_015637619.1 | 6 - 7 | 1204 | 66 | ||
| GenBank mRNA | AK103430.1 | - | 3 - 4 | 209 | 66 | |
More Information
Set Membership: |
|

LOC4338177
Genomic Map