Assay Details
| Gene Symbol: LOC4338177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782132.2 | XP_015637618.1 | 10 - 11 | 1391 | 76 | |
| XM_015782133.2 | XP_015637619.1 | 9 - 10 | 1530 | 76 | ||
| GenBank mRNA | AK103430.1 | - | 6 - 7 | 535 | 76 | |
More Information
Set Membership: |
|

LOC4338177
Genomic Map