Assay Details
| Gene Symbol: LOC4338922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782165.2 | XP_015637651.1 | 6 - 7 | 591 | 103 | |
| XM_015782166.2 | XP_015637652.1 | 6 - 7 | 705 | 103 | ||
More Information
Set Membership: |
|

LOC4338922
Genomic Map