Assay Details
| Gene Symbol: LOC4339019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782432.2 | XP_015637918.1 | 1 - 2 | 144 | 116 | |
| XM_015782433.2 | XP_015637919.1 | 1 - 2 | 145 | 116 | ||
| GenBank mRNA | AK058842.1 | - | 1 - 2 | 124 | 116 | |
| AK066350.1 | - | 1 - 2 | 103 | 116 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015782433.2 transcripts.
Set Membership: |
|

LOC4339019
Genomic Map