Assay Details
| Gene Symbol: LOC4339099 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781842.2 | XP_015637328.1 | 14 - 15 | 2237 | 81 | |
| GenBank mRNA | AK063430.1 | - | 14 - 15 | 2225 | 81 | |
| AK099369.1 | - | 14 - 15 | 2180 | 81 | ||
More Information
Set Membership: |
|

LOC4339099
Genomic Map