Assay Details
| Gene Symbol: LOC4339099 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781842.2 | XP_015637328.1 | 5 - 6 | 879 | 91 | |
| GenBank mRNA | AK063430.1 | - | 5 - 6 | 867 | 91 | |
| AK099369.1 | - | 5 - 6 | 822 | 91 | ||
More Information
Set Membership: |
|

LOC4339099
Genomic Map