Assay Details
| Gene Symbol: LOC4339099 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781842.2 | XP_015637328.1 | 9 - 10 | 1644 | 96 | |
| GenBank mRNA | AK063430.1 | - | 9 - 10 | 1632 | 96 | |
| AK099369.1 | - | 9 - 10 | 1587 | 96 | ||
More Information
Set Membership: |
|

LOC4339099
Genomic Map