Assay Details
| Gene Symbol: LOC9268106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783825.2 | XP_015639311.1 | 1 | 1323 | 118 | |
| GenBank mRNA | AK108158.1 | - | 1 | 1055 | 118 | |
More Information
Set Membership: |
|

LOC9268106
Genomic Map