Assay Details
| Gene Symbol: LOC4339304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783273.2 | XP_015638759.1 | 7 - 8 | 812 | 63 | |
| GenBank mRNA | AK101866.1 | - | 7 - 8 | 818 | 63 | |
More Information
Set Membership: |
|

LOC4339304
Genomic Map