Assay Details
| Gene Symbol: LOC4339457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783720.2 | XP_015639206.1 | 12 - 13 | 1703 | 119 | |
| XM_015783721.1 | XP_015639207.1 | 11 - 12 | 1631 | 119 | ||
| XM_015783722.2 | XP_015639208.1 | 12 - 13 | 1655 | 119 | ||
| GenBank mRNA | AK068572.1 | - | 12 - 13 | 1424 | 119 | |
More Information
Set Membership: |
|

LOC4339457
Genomic Map