Assay Details
| Gene Symbol: LOC4339457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783720.2 | XP_015639206.1 | 15 - 16 | 1972 | 101 | |
| XM_015783721.1 | XP_015639207.1 | 14 - 15 | 1900 | 101 | ||
| XM_015783722.2 | XP_015639208.1 | 15 - 16 | 1924 | 101 | ||
| GenBank mRNA | AK068572.1 | - | 15 - 16 | 1693 | 101 | |
More Information
Set Membership: |
|

LOC4339457
Genomic Map