Assay Details
| Gene Symbol: LOC4339457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783720.2 | XP_015639206.1 | 3 - 4 | 821 | 72 | |
| XM_015783721.1 | XP_015639207.1 | 3 - 4 | 821 | 72 | ||
| XM_015783722.2 | XP_015639208.1 | 3 - 4 | 773 | 72 | ||
| GenBank mRNA | AK068572.1 | - | 3 - 4 | 542 | 72 | |
More Information
Set Membership: |
|

LOC4339457
Genomic Map