Assay Details
| Gene Symbol: LOC4339457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783720.2 | XP_015639206.1 | 7 - 8 | 1154 | 90 | |
| XM_015783721.1 | XP_015639207.1 | 6 - 7 | 1082 | 90 | ||
| XM_015783722.2 | XP_015639208.1 | 7 - 8 | 1106 | 90 | ||
| GenBank mRNA | AK068572.1 | - | 7 - 8 | 875 | 90 | |
More Information
Set Membership: |
|

LOC4339457
Genomic Map