Assay Details
| Gene Symbol: LOC4339750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784568.2 | XP_015640054.1 | 2 - 3 | 580 | 63 | |
| XM_015784569.2 | XP_015640055.1 | 2 - 3 | 581 | 63 | ||
| GenBank mRNA | AK067538.1 | - | 2 - 3 | 348 | 63 | |
More Information
Set Membership: |
|

LOC4339750
Genomic Map