Assay Details
| Gene Symbol: LOC4339750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784568.2 | XP_015640054.1 | 4 - 5 | 826 | 85 | |
| XM_015784569.2 | XP_015640055.1 | 4 - 5 | 827 | 85 | ||
| GenBank mRNA | AK067538.1 | - | 4 - 5 | 594 | 85 | |
More Information
Set Membership: |
|

LOC4339750
Genomic Map