Assay Details
| Gene Symbol: LOC4339750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784568.2 | XP_015640054.1 | 8 - 9 | 1127 | 99 | |
| XM_015784569.2 | XP_015640055.1 | 8 - 9 | 1128 | 99 | ||
| GenBank mRNA | AK067538.1 | - | 8 - 9 | 895 | 99 | |
More Information
Set Membership: |
|

LOC4339750
Genomic Map