Assay Details
| Gene Symbol: LOC4339891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788226.2 | XP_015643712.1 | 8 - 9 | 2738 | 66 | |
| GenBank mRNA | AK069834.1 | - | 8 - 9 | 2739 | 66 | |
| AK071761.1 | - | 8 - 9 | 1650 | 66 | ||
More Information
Set Membership: |
|

LOC4339891
Genomic Map