Assay Details
| Gene Symbol: LOC4339897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK061381.1 | - | 1 - 2 | 303 | 110 | |
| AK103951.1 | - | 1 - 2 | 278 | 110 | ||
| AK104422.1 | - | 1 - 2 | 290 | 110 | ||
| AK104612.1 | - | 1 - 2 | 268 | 110 | ||
More Information
Set Membership: |
|

LOC4339897
Genomic Map